missing translation for 'onlineSavingsMsg'
Learn More

BIX02188, MedChemExpress

Product Code. 16464958
Encompass_Preferred
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Mængde:
5 mg
10 mg
50 mg
100 mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
16464958 10 mg
16437697 5 mg
16431207 50 mg
16415736 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 16464958 missing translation for 'mfr' Medchem Express missing translation for 'supplierNo' HY12055

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress BIX02188 is a potent MEK5-selective inhibitor with an IC50 of 4.3 nM. BIX02188 inhibits ERK5 catalytic activity, with an IC50 of 810 nM.

BIX02188 is a potent MEK5-selective inhibitor with an IC50 of 4.3 nM. BIX02188 inhibits ERK5 catalytic activity, with an IC50 of 810 nM.

Specifications

Kemisk navn eller materiale BIX02188
Fysisk form Solid
CAS 334949-59-6
Farve Yellow
Mængde 10 mg
Renhedsgrad noter Research
Molekylær formel C25H24N4O2
Til brug med (applikation) Cancer-Kinase/protease
Opløselighedsinformation DMSO : ≥ 45 mg/mL (109.10 mM)
SMIL O=C(N)C1=CC(NC2=O)=C(/C2=C(C3=CC=CC=C3)/NC4=CC=CC(CN(C)C)=C4)C=C1.[(Z)]
Molekylvægt (g/mol) 412.48
Formel vægt 412.48
Procent renhed 98.02%
Grad Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.