Få mere at vide
Tocris Bioscience™ BIX NHE1 inhibitor
Potent and selective NHE1 inhibitor
Brand: Tocris Bioscience™ 5512/10
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
BIX NHE1 inhibitor is a potent and selective NHE1 inhibitor (IC50 = 31 nM). Selective for NHE1 over NHE2 and NHE3. Prevents ischemic damage in an ischemia reperfusion injury isolated rat heart model ex vivo. Prevents phenylephrine -induced cardiomyocyte hypertrophy in vitro, and attenuates cardiac hypertrophy and left ventricular dysfunction postinfarction in rats. Orally bioavailable.Tekniske data
| 4-(1-Acetyl-4-piperidinyl)-N-(aminoiminomethyl)-3-(trifluoromethyl)benzamide hydrochloride | |
| 1422252-46-7 | |
| 10 mg |