Få mere at vide
Bisphenol F Monosulfate Sodium Salt, TRC
Artikelnummer. 30137380
Skift visning
Mængde:
50 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30137380 100 mg
17993385 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30137380 Leverandør Toronto Research Chemicals Leverandørnr. TRCB519595100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 54172-90-6
Molekylær formel C13H11NaO5S
Molekylvægt (g/mol) 302.28
Synonym 4-(4-Hydroxybenzyl)phenyl Hydrogen Sulfate Sodium Salt, BPF Monosulfate Sodium Salt;
IUPAC navn [4-[(4-hydroxyphenyl)methyl]phenoxy]sulfonyloxysodium
SMIL Oc1ccc(Cc2ccc(OS(=O)(=O)O[Na])cc2)cc1
Tekniske data
Kemisk navn eller materiale Bisphenol F Monosulfate Sodium Salt
Mængde 100 mg
InChI formel InChI=1S/C13H12O5S.Na/c14-12-5-1-10(2-6-12)9-11-3-7-13(8-4-11)18-19(15,16)17;/h1-8,14H,9H2,(H,15,16,17);/q;+1/p-1
Formel vægt 302.0225