Få mere at vide
Bisphenol A-d8, TRC
Artikelnummer. 30348082
Skift visning
Mængde:
100 mg
Artikelnummer. Mængde
30348082 100 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30348082 Leverandør Toronto Research Chemicals Leverandørnr. TRCB519499100MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Bisphenol A ring-d8
CAS 92739-58-7
Renhedsgrad noter HPLC
Molekylær formel C15 2H8 H8 O2
Mængde 100 mg
Synonym Phen-2,3,5,6-d4-ol, 4,4'-(1-methylethylidene)bis-, 4,4'-(1-Methylethylidene)bis[phen-2,3,5,6-d4-ol], Bisphenol A-d8, Bisphenol A (ring-D8), 2,2-Bis(4-hydroxy-2,3,5,6-tetradeuteriophenyl)propane, 4,4'-Isopropylidenediphenol (ring-D8)
InChI formel InChI=1S/C15H16O2/c1-15(2,11-3-7-13(16)8-4-11)12-5-9-14(17)10-6-12/h3-10,16-17H,1-2H3/i3D,4D,5D,6D,7D,8D,9D,10D
SMIL [2H]c1c([2H])c(c([2H])c([2H])c1O)C(C)(C)c2c([2H])c([2H])c(O)c([2H])c2[2H]
IUPAC navn 2,3,5,6-tetradeuterio-4-[2-(2,3,5,6-tetradeuterio-4-hydroxyphenyl)propan-2-yl]phenol
Molekylvægt (g/mol) 236.34
Formel vægt 236.1652
Procent renhed >95