Få mere at vide
Bisphenol A-D6, TRC
Artikelnummer. 30340485
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30340485 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30340485 Leverandør Toronto Research Chemicals Leverandørnr. TRCB51949710MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Bisphenol A-d6
CAS 86588-58-1
Renhedsgrad noter HPLC
Molekylær formel C15 2H6 H10 O2
Mængde 10 mg
Synonym Phenol, 4,4'-[1-(methyl-d3)ethylidene-2,2,2-d3]bis-, 4,4'-[1-(Methyl-d3)ethylidene-2,2,2-d3]bis[phenol], 2,2-Bis(4-hydroxyphenyl)-1,1,1,3,3,3-hexadeuteriopropane, 4,4'-Isopropylidenediphenol (isopropyl-D6), Bisphenol A (propane-D6), Bisphenol A (dimethyl-D6)
InChI formel InChI=1S/C15H16O2/c1-15(2,11-3-7-13(16)8-4-11)12-5-9-14(17)10-6-12/h3-10,16-17H,1-2H3/i1D3,2D3
SMIL [2H]C([2H])([2H])C(c1ccc(O)cc1)(c2ccc(O)cc2)C([2H])([2H])[2H]
IUPAC navn 4-[1,1,1,3,3,3-hexadeuterio-2-(4-hydroxyphenyl)propan-2-yl]phenol
Molekylvægt (g/mol) 234.32
Formel vægt 234.1527
Procent renhed >95