Få mere at vide
Bisphenol A-d6 Beta-D-Glucuronide, TRC
Artikelnummer. 30340084
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30340084 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30340084 Leverandør Toronto Research Chemicals Leverandørnr. TRCB5195121MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Bisphenol A beta-D-Monoglucuronide dimethyl-d6
CAS 1610029-53-2
Molekylær formel C21 2H6 H18 O8
Mængde 1 mg
Synonym β-D-Glucopyranosiduronic acid, 4-[1-(4-hydroxyphenyl)-1-(methyl-d3)ethyl-2,2,2-d3]phenyl, 4-[1-(4-Hydroxyphenyl)-1-(methyl-d3)ethyl-2,2,2-d3]phenyl β-D-glucopyranosiduronic acid, Bisphenol A β-D-Glucuronide (dimethyl-D6), Bisphenol A β-D-Monoglucuronide (dimethyl-D6)
InChI formel InChI=1S/C21H24O8/c1-21(2,11-3-7-13(22)8-4-11)12-5-9-14(10-6-12)28-20-17(25)15(23)16(24)18(29-20)19(26)27/h3-10,15-18,20,22-25H,1-2H3,(H,26,27)/t15-,16-,17+,18-,20+/m0/s1/i1D3,2D3
SMIL [2H]C([2H])([2H])C(c1ccc(O)cc1)(c2ccc(O[C@@H]3O[C@@H]([C@@H](O)[C@H](O)[C@H]3O)C(=O)O)cc2)C([2H])([2H])[2H]
IUPAC navn (2S,3S,4S,5R,6S)-6-[4-[1,1,1,3,3,3-hexadeuterio-2-(4-hydroxyphenyl)propan-2-yl]phenoxy]-3,4,5-trihydroxyoxane-2-carboxylic acid
Molekylvægt (g/mol) 410.45
Formel vægt 410.1848