Få mere at vide
Bisphenol A-3,3',5,5'-d4, TRC
Artikelnummer. 30344067
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30344067 5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30344067 Leverandør Toronto Research Chemicals Leverandørnr. TRCB5195085MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Bisphenol A-3,3',5,5'-d4
CAS 347841-41-2
Molekylær formel C15 D4 H12 O2
Mængde 5 mg
Synonym Phen-2,6-d2-ol, 4,4'-(1-methylethylidene)bis- (9CI), Bisphenol A-3,3',5,5'-d4, Bisphenol A-3,3',5,5'-D4, 4,4'-(1-Methylethylidene)bisphen-2,6-d2-ol
InChI formel InChI=1S/C15H16O2/c1-15(2,11-3-7-13(16)8-4-11)12-5-9-14(17)10-6-12/h3-10,16-17H,1-2H3/i7D,8D,9D,10D
SMIL [2H]c1cc(cc([2H])c1O)C(C)(C)c2cc([2H])c(O)c([2H])c2
IUPAC navn 2,6-dideuterio-4-[2-(3,5-dideuterio-4-hydroxyphenyl)propan-2-yl]phenol
Molekylvægt (g/mol) 232.311
Formel vægt 232.1401