Få mere at vide
Bisoprolol-d5, TRC
Artikelnummer. 30348237
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30348237 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30348237 Leverandør Toronto Research Chemicals Leverandørnr. TRCB51050210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Bisoprolol-d5
CAS 1189881-87-5
Molekylær formel C18H26D5NO4
Mængde 10 mg
Synonym 1-[4-[[2-(1-Methylethoxy)ethoxy]methyl]phenoxy]-3-[(1-methylethyl)amino]-2-propanol-d5
InChI formel InChI=1S/C18H31NO4/c1-14(2)19-11-17(20)13-23-18-7-5-16(6-8-18)12-21-9-10-22-15(3)4/h5-8,14-15,17,19-20H,9-13H2,1-4H3/i11D2,13D2,17D
SMIL CC(C)OCCOCC1=CC=C(OC([2H])([2H])C(O)([2H])C([2H])([2H])NC(C)C)C=C1
IUPAC navn 1,1,2,3,3-pentadeuterio-1-(propan-2-ylamino)-3-[4-(2-propan-2-yloxyethoxymethyl)phenoxy]propan-2-ol
Molekylvægt (g/mol) 330.47
Formel vægt 330.2567