missing translation for 'onlineSavingsMsg'
Få mere at vide

Bismuth Tripotassium Dicitrate (>85%), TRC

Artikelnummer. 30328829
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30328829 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30328829 Supplier Toronto Research Chemicals Supplier No. TRCB4973101G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale Bismuth Tripotassium Dicitrate
CAS 57644-54-9
Molekylær formel C12H10BiK3O14
Mængde 1 g
Synonym Bicit HP, Bismuth Subcitrate, Bismuth Tripotassium Dicitrate, CBS, Colloidal Bismuth Subcitrate, De-Nol, De-Noltab, Duosol, Duosol (ulcer treatment), Gastrodenol, Q-Ulcer, Tripotassium Dicitratobismuthate, Trymo, Ulcerone
InChI formel InChI=1S/2C6H8O7.Bi.3K/c2*7-3(8)1-6(13,5(11)12)2-4(9)10;;;;/h2*13H,1-2H2,(H,7,8)(H,9,10)(H,11,12);;;;/q;;+3;3*+1/p-6
SMIL [K+].[K+].[K+].[Bi+3].OC(CC(=O)[O-])(CC(=O)[O-])C(=O)[O-].OC(CC(=O)[O-])(CC(=O)[O-])C(=O)[O-]
IUPAC navn bismuth;tripotassium;2-hydroxypropane-1,2,3-tricarboxylate
Molekylvægt (g/mol) 704.47
Formel vægt 703.8786
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.