Få mere at vide
Bis-TRIS-d19, CDN
Artikelnummer. 30309680
Skift visning
Mængde:
0.25 g
0.5 g
Pakningsstørrelse:
0.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30309680 0.25 g -
30319346 0.5 g 0.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30309680 Leverandør CDN Leverandørnr. CDND64570.25G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Bis-TRIS-d19
CAS 352534-93-1
Molekylær formel C8 D19 N O5
Mængde 0.25 g
Synonym 1,3-Propane-1,1,3,3-d4-diol-d2, 2-[bis[2-(hydroxy-d)ethyl-1,1,2,2-d4]amino]-2-(hydroxy-d-methyl-d2)- (9 CI)
InChI formel InChI=1 S/C8H19NO5/c10-3-1-9(2-4-11)8(5-12,6-13)7-14/h10-14 H,1-7H2/i1D2,2D2,3D2,4D2,5D2,6D2,7D2,10 D,11 D,12 D,13 D,14 D
SMIL [2 H]OC([2 H])([2 H])C([2 H])([2 H])N(C([2 H])([2 H])C([2 H])([2 H])O[2 H])C(C([2 H])([2 H])O[2 H])(C([2 H])([2 H])O[2 H])C([2 H])([2 H])O[2 H]
IUPAC navn 1,1,3,3-tetradeuterio-1,3-dideuteriooxy-2-[dideuterio(deuteriooxy)methyl]-N,N-bis(1,1,2,2-tetradeuterio-2-deuteriooxyethyl)propan-2-amine
Molekylvægt (g/mol) 228.357
Formel vægt 228.246 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity