missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals BIS-TRIS, 0.2M buffer soln., pH 6.5

Catalog Number p-7057381
Click to view available options
Quantity:
100 mL
250 mL
This item is not returnable. View return policy
6976-37-0
C8H19NO5
209.242
MFCD00002853
OWMVSZAMULFTJU-UHFFFAOYSA-N
81462
CHEBI:41250
2-[bis(2-hydroxyethyl)amino]-2-(hydroxymethyl)propane-1,3-diol
C(CO)N(CCO)C(CO)(CO)CO
This item is not returnable. View return policy
6976-37-0
C8H19NO5
209.242
MFCD00002853
OWMVSZAMULFTJU-UHFFFAOYSA-N
81462
CHEBI:41250
2-[bis(2-hydroxyethyl)amino]-2-(hydroxymethyl)propane-1,3-diol
C(CO)N(CCO)C(CO)(CO)CO

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Colorless
Concentration 0.2 M
pH 6.5
Physical Form Liquid
Quantity 100 mL
Odor Odorless
Chemical Name or Material BIS-TRIS

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'