missing translation for 'onlineSavingsMsg'
Learn More

Bis(triethylphosphine)palladium(II) chloride, ACROS Organics™

294.00 DKK - 885.00 DKK

Chemical Identifiers

CAS 28425-04-9
Molecular Formula C12H30Cl2P2Pd
Molecular Weight (g/mol) 413.64
MDL Number MFCD00191831
InChI Key ULYNIEUXPCUIEL-UHFFFAOYSA-L
Synonym dichlorobis triethylphosphine palladium ii, bis triethylphosphine palladium dichloride, trans-dichlorobis triethylphosphine palladium ii, bis triethylphosphine palladium ii chloride, pdcl2 pet3 2, palladium,dichlorobis triethylphosphine, dichloropalladium-triethylphosphane 1/2, bis triethylphosphine dichloridopalladium ii
PubChem CID 11144124
IUPAC Name dichloropalladium;triethylphosphane
SMILES CCP(CC)CC.CCP(CC)CC.Cl[Pd]Cl
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10722104
SDS View Documents
Acros Organics
377972500
250mg Glass bottle
294.00 DKK
250mg
View alternative products
Save Up to 
»
Estimated Shipment
26-04-2021
Please to purchase this item. Need a web account? Register with us today!
10443175
SDS View Documents
Acros Organics
377970010
1g Glass bottle
885.00 DKK
1g
View alternative products
Save Up to 
»
In stock, ready to ship: 1 Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

28425-04-9
413.64
ULYNIEUXPCUIEL-UHFFFAOYSA-L
11144124
CCP(CC)CC.CCP(CC)CC.Cl[Pd]Cl
C12H30Cl2P2Pd
MFCD00191831
dichlorobis triethylphosphine palladium ii, bis triethylphosphine palladium dichloride, trans-dichlorobis triethylphosphine palladium ii, bis triethylphosphine palladium ii chloride, pdcl2 pet3 2, palladium,dichlorobis triethylphosphine, dichloropalladium-triethylphosphane 1/2, bis triethylphosphine dichloridopalladium ii
dichloropalladium;triethylphosphane

Specifications

Bis(triethylphosphine)palladium(II) chloride
Glass bottle
139°C to 142°C
28425-04-9
MFCD00191831
Solubility in water: insoluble
CCP(CC)CC.CCP(CC)CC.Cl[Pd]Cl
413.64
413.64
Authentic
Yellow
250mg
C12H30Cl2P2Pd
dichlorobis triethylphosphine palladium ii, bis triethylphosphine palladium dichloride, trans-dichlorobis triethylphosphine palladium ii, bis triethylphosphine palladium ii chloride, pdcl2 pet3 2, palladium,dichlorobis triethylphosphine, dichloropalladium-triethylphosphane 1/2, bis triethylphosphine dichloridopalladium ii
ULYNIEUXPCUIEL-UHFFFAOYSA-L
dichloropalladium;triethylphosphane
11144124
Powder
Safety and Handling

Safety and Handling

Product Identifier
  • Bis(triethylphosphine)palladium(II) chloride

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H332-Harmful if inhaled.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS