Få mere at vide
Bis(pentamethylcyclopentadienyl)manganese, 98%, Thermo Scientific™
Artikelnummer. 10530722 Shop alle Thermo Scientific Chemicals produkter
Skift visning
Mængde:
1 g
Pakningsstørrelse:
1g
Artikelnummer. Mængde unitSize
10530722 1 g 1g
1 options
Artikelnummer. 10530722 Leverandør Thermo Scientific Acros Leverandørnr. 353260010
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 67506-86-9
Molekylær formel C20H30Mn
MDL nummer MFCD00058844
Tekniske data
Kemisk navn eller materiale Bis(pentamethylcyclopentadienyl)manganese
Farve Red to Brown
Infrarødt spektrum Authentic
Analyseprocentområde 98%
Emballage Glass bottle
Mængde 1 g
Formel vægt 325.39
Fysisk form Crystalline Powder