Få mere at vide
Bis(N,N-dibutylamine)-N,N’-disulfide-d36, TRC
Artikelnummer. 30341839
Skift visning
Mængde:
2.5 mg
Pakningsstørrelse:
2.5mg
Artikelnummer. Mængde unitSize
30341839 2.5 mg 2.5mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30341839 Leverandør Toronto Research Chemicals Leverandørnr. TRCB4261622.5MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Bis(N,N-dibutylamine)-N,N'-disulfide-d36
CAS 1216478-97-5
Molekylær formel C16D36N2S2
Mængde 2.5 mg
Synonym N,N’-Dithiobis[N-butyl]-1-butanamine-d36, N,N’-Dithiobis-dibutylamine-d36
InChI formel InChI=1S/C16H36N2S2/c1-5-9-13-17(14-10-6-2)19-20-18(15-11-7-3)16-12-8-4/h5-16H2,1-4H3/i1D3,2D3,3D3,4D3,5D2,6D2,7D2,8D2,9D2,10D2,11D2,12D2,13D2,14D2,15D2,16D2
SMIL [2H]C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])N(SSN(C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])[2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])[2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])[2H]
IUPAC navn N-[[bis(1,1,2,2,3,3,4,4,4-nonadeuteriobutyl)amino]disulfanyl]-1,1,2,2,3,3,4,4,4-nonadeuterio-N-(1,1,2,2,3,3,4,4,4-nonadeuteriobutyl)butan-1-amine
Molekylvægt (g/mol) 356.82
Formel vægt 356.458