Få mere at vide
Bis-L-phenylalanylcyclohexanedimethanol, TRC
Artikelnummer. 30342560
Skift visning
Mængde:
500 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30342560 1 g
30329697 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30342560 Leverandør Toronto Research Chemicals Leverandørnr. TRCB5212501G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Bis-L-phenylalanylcyclohexanedimethanol
CAS 205654-87-1
Molekylær formel C26 H34 N2 O4
Mængde 1 g
InChI formel InChI=1S/C26H34N2O4/c27-23(15-19-7-3-1-4-8-19)25(29)31-17-21-11-13-22(14-12-21)18-32-26(30)24(28)16-20-9-5-2-6-10-20/h1-10,21-24H,11-18,27-28H2/t21?,22?,23-,24-/m0/s1
SMIL N[C@@H](Cc1ccccc1)C(=O)OCC2CCC(COC(=O)[C@@H](N)Cc3ccccc3)CC2
IUPAC navn [4-[[(2S)-2-amino-3-phenylpropanoyl]oxymethyl]cyclohexyl]methyl (2S)-2-amino-3-phenylpropanoate
Molekylvægt (g/mol) 438.56
Formel vægt 438.2519