Få mere at vide
Bis(heptadecafluorooctyl)phosphinic Acid Sodium Salt, TRC
Artikelnummer. 30134084
Skift visning
Mængde:
50 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30134084 500 mg
17948779 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30134084 Leverandør Toronto Research Chemicals Leverandørnr. TRCB443750500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 500776-69-2
Molekylær formel C16F34NaO2P
Molekylvægt (g/mol) 924.08
Synonym Bis(perfluorooctyl)phosphinic Acid, Bis(heptadecafluorooctyl)phosphinic Acid Sodium Salt, P,P-Bis(1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-heptadecafluorooctyl)phosphinic Acid Sodium Salt;
IUPAC navn sodium;bis(1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-heptadecafluorooctyl)phosphinate
SMIL O=P(C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F)(C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F)O[Na]
Tekniske data
Kemisk navn eller materiale Bis(heptadecafluorooctyl)phosphinic Acid Sodium Salt
Mængde 500 mg
InChI formel InChI=1S/C16HF34O2P.Na/c17-1(18,5(25,26)9(33,34)13(41,42)43)3(21,22)7(29,30)11(37,38)15(47,48)53(51,52)16(49,50)12(39,40)8(31,32)4(23,24)2(19,20)6(27,28)10(35,36)14(44,45)46;/h(H,51,52);/q;+1/p-1
Formel vægt 923.8991