missing translation for 'onlineSavingsMsg'
Learn More

Bis(diphenylphosphinomethyl)phenylphosphine oxide, 95%, Thermo Scientific™

Catalog Number p-7032881
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
21851-89-8
C32H29O3P3
554.50
MFCD00482218
GPDBOGDYDXUTRD-UHFFFAOYSA-N
bis diphenylphosphinomethyl phenylphosphine oxide, diphenylphosphanyl methyl phenyl phosphoryl methyl diphenylphosphane, 1,1,3,5,5-pentaphenyl-1,3,5-triphosphapentane trioxide
[({[(diphenylphosphoroso)methyl](phenyl)phosphoryl}methyl)(phenyl)phosphoroso]benzene
O=P(CP(=O)(C1=CC=CC=C1)C1=CC=CC=C1)(CP(=O)(C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1
This item is not returnable. View return policy
21851-89-8
C32H29O3P3
554.50
MFCD00482218
GPDBOGDYDXUTRD-UHFFFAOYSA-N
bis diphenylphosphinomethyl phenylphosphine oxide, diphenylphosphanyl methyl phenyl phosphoryl methyl diphenylphosphane, 1,1,3,5,5-pentaphenyl-1,3,5-triphosphapentane trioxide
[({[(diphenylphosphoroso)methyl](phenyl)phosphoryl}methyl)(phenyl)phosphoroso]benzene
O=P(CP(=O)(C1=CC=CC=C1)C1=CC=CC=C1)(CP(=O)(C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 554.59
Percent Purity 95%
Chemical Name or Material Bis(diphenylphosphinomethyl)phenylphosphine oxide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'