Få mere at vide
Bis(diphenylmethyl)disulfide, Mikromol™
Beskrivelse
Bis(diphenylmethyl)disulfide
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Bis(diphenylmethyl)disulfide |
| CAS | 1726-02-9 |
| Analyt- eller komponentnavne | Bis(diphenylmethyl)disulfide |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C26 H22 S2 |
| Certificeringer/compliance | ISO 17025 |
| Synonym | Bis(diphenylmethyl)disulfide, Modafinil Related Compound E |
| InChI formel | InChI=1S/C26H22S2/c1-5-13-21(14-6-1)25(22-15-7-2-8-16-22)27-28-26(23-17-9-3-10-18-23)24-19-11-4-12-20-24/h1-20,25-26H |
| SMIL | S(SC(c1ccccc1)c2ccccc2)C(c3ccccc3)c4ccccc4 |
| IUPAC navn | [(benzhydryldisulfanyl)-phenylmethyl]benzene |
| Vis mere |