missing translation for 'onlineSavingsMsg'
Få mere at vide

Bis(diethylamino)chlorophosphine, 97%, Thermo Scientific™

Artikelnummer. 10073870 Shop alle Thermo Scientific Chemicals produkter
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
1 g
Emballage:
Glass bottle
Pakningsstørrelse:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde Emballage unitSize
10073870 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 10073870 Leverandør Thermo Scientific Acros Leverandørnr. 317360010
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter

Denne vare kan ikke returneres. Se returpolicy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale Bis(diethylamino)chlorophosphine
Navn note 97%
CAS 685-83-6
CAS min % 96.0
CAS Max % 100.0
Farve Colorless to Yellow
Tæthed 1.0000g/mL
Kogepunkt 55.0°C to 57.0°C (0.2 mmHg)
Flammepunkt 101°C
Infrarødt spektrum Authentic
Analyseprocentområde 96% min. (ex Cl) (Argentometry)
Emballage Glass bottle
Molekylær formel C8H20ClN2P
Lineær formel [(C2H5)2N]2PCl
Brydningsindeks 1.4885 to 1.4905
MDL nummer MFCD00015493
Mængde 1 g
Specifik vægtfylde 1
Synonym bis diethylamino chlorophosphine, bis diethylamino phosphorchloridite, n,n,n',n'-tetraethylphosphorodiamidous chloride, bdapcd, chloro diethylamino phosphanyl diethylamine, pubchem6475, phosphorodiamidous chloride, tetraethyl, chlorobis diethylamino phosphine, bisdiethylamino-chloro-phosphine, chloro-bis diethylamino-phosphine
Opløselighedsinformation Solubility in water: reacts with water
InChI nøgle JVEHJSIFWIIFHM-UHFFFAOYSA-N
SMIL CCN(CC)P(N(CC)CC)Cl
IUPAC navn N-[chloro(diethylamino)phosphanyl]-N-ethylethanamine
Molekylvægt (g/mol) 210.69
PubChem CID 2733351
Formel vægt 210.69
Procent renhed 97%
Fysisk form Liquid
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.