missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bis(cyclopentadienyl)zirconium chloride hydride, 95%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent
Specifications
CAS Min % | 94.0 |
CAS Max % | 100.0 |
Melting Point | >300.0°C |
Color | White to Beige |
Infrared Spectrum | Authentic |
Assay Percent Range | 94% min. (Argentometry) |
Packaging | Glass bottle |
Quantity | 5 g |
Merck Index | 15, 8533 |
Solubility Information | Solubility in water: reacts with water. |
Show More |
Safety and Handling
Product Identifier
- Bis(cyclopentadienyl)zirconium chloride hydride
- Danger
- Serious eye damage/eye irritation Category 1
- Flammable solid Category 2
- Skin corrosion/irritation Category 1 B
- Substance which\, in contact with water\, emits flammable gases Category 2
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H261-In contact with water releases flammable gases.
- H228-Flammable solid.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'