Få mere at vide
Bis[a,a-bis(trifluoromethyl)benzenemethanolato]diphenylsulfur, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Bis[a,a-bis(trifluoromethyl)benzenemethanolato]diphenylsulfur |
| CAS | 32133-82-7 |
| Molekylær formel | C30 H20 F12 O2 S |
| Mængde | 1 g |
| Synonym | Sulfur, bis[[α,α-bis(trifluoromethyl)benzyl]oxy]diphenyl- (8CI), Sulfur, bis[α,α-bis(trifluoromethyl)benzenemethanolato]diphenyl-, (T4)-, (T-4)-Bis[α,α-bis(trifluoromethyl)benzenemethanolato-κO]diphenylsulfur, Benzenemethanol, α,α-bis(trifluoromethyl)-, sulfur complex, Bis[bis(trifluoromethyl)phenylmethoxy]diphenylsulfurane, Diphenylbis(1,1,1,3,3,3-hexafluoro-2-phenyl-2-propoxy)sulfurane, Martin sulfurane, Martin's sulfurane |
| InChI formel | InChI=1S/C30H20F12O2S/c31-27(32,33)25(28(34,35)36,21-13-5-1-6-14-21)43-45(23-17-9-3-10-18-23,24-19-11-4-12-20-24)44-26(29(37,38)39,30(40,41)42)22-15-7-2-8-16-22/h1-20H |
| SMIL | FC(F)(F)C(OS(OC(c1ccccc1)(C(F)(F)F)C(F)(F)F)(c2ccccc2)c3ccccc3)(c4ccccc4)C(F)(F)F |
| IUPAC navn | bis[(1,1,1,3,3,3-hexafluoro-2-phenylpropan-2-yl)oxy]-diphenyl-λ^{4}-sulfane |
| Molekylvægt (g/mol) | 672.524 |
| Formel vægt | 672.099 |