Få mere at vide
Bis(7-methyl-1-octyl) Phthalate-3,4,5,6-d4, CDN
Artikelnummer. 30323504
Skift visning
Mængde:
0.05 g
0.1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30323504 0.05 g
30314766 0.1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30323504 Leverandør CDN Leverandørnr. CDND74470.05G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Phthalic Acid Di-isononyl Ester (3,4,5,6)-D4
CAS 1332965-90-8
Molekylær formel C26 2H4 H38 O4
Mængde 0.05 g
Synonym 1,2-Benzene-3,4,5,6-d4-dicarboxylic acid, 1,2-bis(7-methyloctyl) ester, Diisononyl Phthalate-d4, Phthalic Acid Di-isononyl Ester (3,4,5,6)-D4
InChI formel InChI=1 S/C26H42O4/c1-21(2)15-9-5-7-13-19-29-25(27)23-17-11-12-18-24(23)26(28)30-20-14-8-6-10-16-22(3)4/h11-12,17-18,21-22 H,5-10,13-16,19-20H2,1-4H3/i11D,12 D,17 D,18 D
SMIL [2 H]c1c([2 H])c([2 H])c(C(=O)OCCCCCCC(C)C)c(C(=O)OCCCCCCC(C)C)c1[2 H]
IUPAC navn bis(7-methyloctyl) 3,4,5,6-tetradeuteriobenzene-1,2-dicarboxylate
Molekylvægt (g/mol) 422.63
Formel vægt 422.3334 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity