missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bis(4-nitrophenyl)phosphoric Acid, TRC
Specifications
Specifications
| Kemisk navn eller materiale | Bis(4-nitrophenyl)phosphoric Acid |
| CAS | 645-15-8 |
| Molekylær formel | C12 H9 N2 O8 P |
| Mængde | 1 g |
| Synonym | Ribavirin Impurity 29 (Bis(p-nitrophenyl) phosphate) |
| InChI formel | InChI=1S/C12H9N2O8P/c15-13(16)9-1-5-11(6-2-9)21-23(19,20)22-12-7-3-10(4-8-12)14(17)18/h1-8H,(H,19,20) |
| SMIL | OP(=O)(Oc1ccc(cc1)[N+](=O)[O-])Oc2ccc(cc2)[N+](=O)[O-] |
| IUPAC navn | bis(4-nitrophenyl) hydrogen phosphate |
| Molekylvægt (g/mol) | 340.18 |
| Formel vægt | 340.0097 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?