missing translation for 'onlineSavingsMsg'
Learn More

Bis(4-methoxyphenyl) carbonate, 97%, Thermo Scientific™

Catalog Number 11492978
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
5676-71-1
C15H14O5
274.272
MFCD00798557
PVBTWSPZTNYNHG-UHFFFAOYSA-N
bis 4-methoxyphenyl carbonate, 4-methoxyphenyl 4-methoxyphenoxy formate, acmc-1arso, 4-methoxyphenyl carbonate
3793869
bis(4-methoxyphenyl) carbonate
COC1=CC=C(C=C1)OC(=O)OC2=CC=C(C=C2)OC
This item is not returnable. View return policy
5676-71-1
C15H14O5
274.272
MFCD00798557
PVBTWSPZTNYNHG-UHFFFAOYSA-N
bis 4-methoxyphenyl carbonate, 4-methoxyphenyl 4-methoxyphenoxy formate, acmc-1arso, 4-methoxyphenyl carbonate
3793869
bis(4-methoxyphenyl) carbonate
COC1=CC=C(C=C1)OC(=O)OC2=CC=C(C=C2)OC

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 86°C to 88°C
Quantity 10 g
Formula Weight 274.28
Percent Purity 97%
Chemical Name or Material Bis(4-methoxyphenyl) carbonate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'