missing translation for 'onlineSavingsMsg'
Learn More

Bis(4-hydroxy-3,5-dimethylphenyl) Sulfone, TRC

Product Code. 30346391
Change view
Click to view available options
Mængde:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30346391 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30346391 Supplier Toronto Research Chemicals Supplier No. TRCB443878100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale Bis(4-hydroxy-3,5-dimethylphenyl) Sulfone
CAS 13288-70-5
Molekylær formel C16H18O4S
Mængde 100 mg
Synonym 4,4'-Sulfonylbis[2,6-dimethylphenol], 2,2',6,6'-Tetramethyl-4,4'-sulfonyldiphenol, 4,4'-Dihydroxy-3,3',5,5'-tetramethyldiphenyl Sulfone, 4,4'-Dihydroxy-3,3',5,5'-tetramethylphenyl Sulfone, BS-TM, Bis(3,5-dimethyl-4-hydroxyphenyl) Sulfone, Bis(4-hydroxy-3,5-dimethylphenyl) Sulfone, TX 0902
InChI formel InChI=1S/C16H18O4S/c1-9-5-13(6-10(2)15(9)17)21(19,20)14-7-11(3)16(18)12(4)8-14/h5-8,17-18H,1-4H3
SMIL Cc1cc(cc(C)c1O)S(=O)(=O)c2cc(C)c(O)c(C)c2
IUPAC navn 4-(4-hydroxy-3,5-dimethylphenyl)sulfonyl-2,6-dimethylphenol
Molekylvægt (g/mol) 306.38
Formel vægt 306.0926
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.