Få mere at vide
Bis(4-cyanophenyl)methanol-d4, TRC
Artikelnummer. 30329322
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30329322 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30329322 Leverandør Toronto Research Chemicals Leverandørnr. TRCB42285210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Bis(4-cyanophenyl)methanol-d4
CAS 1361326-42-2
Molekylær formel C15H6D4N2O
Mængde 10 mg
Synonym 4,4'-(Hydroxymethylene)bisbenzonitrile-d4, Bis(p-cyanophenyl)methanol-d4, CGP 44645-d4
InChI formel InChI=1S/C15H10N2O/c16-9-11-1-5-13(6-2-11)15(18)14-7-3-12(10-17)4-8-14/h1-8,15,18H/i1D,2D,5D,6D
SMIL [2H]c1c([2H])c(C(O)c2ccc(cc2)C#N)c([2H])c([2H])c1C#N
IUPAC navn 4-[(4-cyanophenyl)-hydroxymethyl]-2,3,5,6-tetradeuteriobenzonitrile
Molekylvægt (g/mol) 238.28
Formel vægt 238.1044