Få mere at vide
Bis[2-(perfluorooctyl)ethyl] phosphate, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Bis(1H,1H,2H,2H-Perfluorodecyl) phosphate |
| CAS | 678-41-1 |
| Renhedsgrad noter | HPLC |
| Molekylær formel | C20 H9 F34 O4 P |
| Mængde | 250 mg |
| Synonym | Bis[2-(perfluorooctyl)ethyl] Phosphate, 1-Decanol, 3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluoro-, hydrogen phosphate (7CI,8CI,9CI), Bis(3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodecyl) hydrogen phosphate, 8:2 Polyfluoroalkylphosphoric acid diester, 1-Decanol, 3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluoro-, 1,1'-(hydrogen phosphate), Bis(1H,1H,2H,2H-Perfluorodecyl) Phosphate |
| InChI formel | InChI=1S/C20H9F34O4P/c21-5(22,7(25,26)9(29,30)11(33,34)13(37,38)15(41,42)17(45,46)19(49,50)51)1-3-57-59(55,56)58-4-2-6(23,24)8(27,28)10(31,32)12(35,36)14(39,40)16(43,44)18(47,48)20(52,53)54/h1-4H2,(H,55,56) |
| SMIL | OP(=O)(OCCC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F)OCCC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F |
| IUPAC navn | bis(3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodecyl) hydrogen phosphate |
| Molekylvægt (g/mol) | 990.2 |
| Vis mere |