missing translation for 'onlineSavingsMsg'
Learn More

Bis(2-methoxyethyl) ether, 99+%, Extra Dry over Molecular Sieve, AcroSeal™, Thermo Scientific Chemicals

Catalog Number p-7077294
Click to view available options
Quantity:
100 mL
1 L
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
111-96-6
C6H14O3
134.18
SBZXBUIDTXKZTM-UHFFFAOYSA-N
diglyme, bis 2-methoxyethyl ether, 2-methoxyethyl ether, 1-methoxy-2-2-methoxyethoxy ethane, diethylene glycol dimethyl ether, poly-solv, di 2-methoxyethyl ether, dimethyl carbitol, 2,5,8-trioxanonane
8150
CHEBI:46784
1-methoxy-2-(2-methoxyethoxy)ethane
COCCOCCOC
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
111-96-6
C6H14O3
134.18
SBZXBUIDTXKZTM-UHFFFAOYSA-N
diglyme, bis 2-methoxyethyl ether, 2-methoxyethyl ether, 1-methoxy-2-2-methoxyethoxy ethane, diethylene glycol dimethyl ether, poly-solv, di 2-methoxyethyl ether, dimethyl carbitol, 2,5,8-trioxanonane
8150
CHEBI:46784
1-methoxy-2-(2-methoxyethoxy)ethane
COCCOCCOC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -64.0°C
Density 0.9370g/mL
Boiling Point 162.0°C
Flash Point 51°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Linear Formula (CH3OCH2CH2)2O
Refractive Index 1.4070 to 1.4090
Quantity 100 mL
Merck Index 15,3184
Specific Gravity 0.937
Solubility Information Solubility in water: miscible. Other solubilities: miscible with alcohol, ether, hydrocarbon solvents
Viscosity 2 mPa.s (20°C)
Formula Weight 134.18
Percent Purity 99+%
Grade Extra Dry over Molecular Sieve
Chemical Name or Material Bis(2-methoxyethyl) ether
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Methoxyethyl ether
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Reproductive toxicity Category 1B
Hazard Statement
  • H360FD-May damage fertility. May damage the unborn child.
  • H226-Flammable liquid and vapour.
  • EUH019-May form explosive peroxides.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'