Få mere at vide
Bis(2-methoxyethyl) 2,6-Dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate, Mikromol™
Artikelnummer. 30052284
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30052284 25 mg
30051271 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30052284 Leverandør Mikromol Leverandørnr. MM0416.030025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Bis(2-methoxyethyl) 2,6-Dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Bis(2-methoxyethyl) 2,6-Dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate
CAS 70172-96-2
Analyt- eller komponentnavne Bis(2-methoxyethyl) 2,6-Dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate
Forsendelsestilstand Room Temperature
Molekylær formel C21 H26 N2 O8
Certificeringer/compliance ISO 9001
Synonym Nimodipine Imp. C (EP), Nimodipine USP Related Compound B, Nimodipine USP RC B, Bis(2-methoxyethyl) 2,6-Dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate, Nimodipine Related Compound B, Nimodipine Impurity C
InChI formel InChI=1S/C21H26N2O8/c1-13-17(20(24)30-10-8-28-3)19(15-6-5-7-16(12-15)23(26)27)18(14(2)22-13)21(25)31-11-9-29-4/h5-7,12,19,22H,8-11H2,1-4H3
SMIL COCCOC(=O)C1=C(C)NC(=C(C1c2cccc(c2)[N+](=O)[O-])C(=O)OCCOC)C
IUPAC navn bis(2-methoxyethyl) 2,6-dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate
Molekylvægt (g/mol) 434.44
Mængde 25 mg
Formel vægt 434.1689 g/mol