Få mere at vide
Bis(2-hydroxyethyl)-1,1,2,2-d4-amine, CDN
Artikelnummer. 30317595
Skift visning
Mængde:
0.05 g
0.1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30317595 0.1 g
30317292 0.05 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30317595 Leverandør CDN Leverandørnr. CDND53070.1G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Diethanolamine-D4 (2-hydroxyethyl-1,1,2,2-D4)
CAS 352431-07-3
Molekylær formel C4 D4 H7 N O2
Mængde 0.1 g
Synonym Ethan-1,1,2,2-d4-ol, 2-[(2-hydroxyethyl)amino]- (9 CI, ACI), 2-[(2-Hydroxyethyl)amino]ethan-1,1,2,2-d4-ol (ACI), Bis(2-hydroxyethyl)-1,1,2,2-d4-amine, Bis(2-hydroxyethyl-1,1,2,2-d4)amine, Diethanolamine-d4, Diethanolamine-D4 (2-hydroxyethyl-1,1,2,2-D4)
InChI formel InChI=1 S/C4H11NO2/c6-3-1-5-2-4-7/h5-7 H,1-4H2/i1D2,3D2
SMIL [2 H]C([2 H])(O)C([2 H])([2 H])NCCO
IUPAC navn 1,1,2,2-tetradeuterio-2-(2-hydroxyethylamino)ethanol
Molekylvægt (g/mol) 109.1603
Formel vægt 109.1041 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity