Få mere at vide
Bis[(±)-2-ethylhexyl] Phthalate-3,4,5,6-d4, CDN
Artikelnummer. 30322628
Skift visning
Mængde:
0.1 g
0.25 g
Pakningsstørrelse:
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30322628 0.25 g -
30310721 0.1 g 0.1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30322628 Leverandør CDN Leverandørnr. CDND23060.25G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Phthalic acid, bis-2-ethylhexyl ester D4
CAS 93951-87-2
Molekylær formel C24 2H4 H34 O4
Mængde 0.25 g
InChI formel InChI=1 S/C24H38O4/c1-5-9-13-19(7-3)17-27-23(25)21-15-11-12-16-22(21)24(26)28-18-20(8-4)14-10-6-2/h11-12,15-16,19-20 H,5-10,13-14,17-18H2,1-4H3/i11D,12 D,15 D,16 D
SMIL [2 H]c1c([2 H])c([2 H])c(C(=O)OCC(CC)CCCC)c(C(=O)OCC(CC)CCCC)c1[2 H]
IUPAC navn bis(2-ethylhexyl) 3,4,5,6-tetradeuteriobenzene-1,2-dicarboxylate
Molekylvægt (g/mol) 394.58
Formel vægt 394.3021 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity