missing translation for 'onlineSavingsMsg'
Få mere at vide

Bis(2-ethylhexyl) Phosphate-d34, TRC

Artikelnummer. 30339928
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30339928 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30339928 Leverandør Toronto Research Chemicals Leverandørnr. TRCB43386725MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale Bis(2-ethylhexyl) Phosphate-d34
CAS 1773493-20-1
Renhedsgrad noter HPLC
Molekylær formel C16 D34 O4 P
Mængde 25 mg
Synonym 2-Ethyl-1-hexanol Hydrogen Phosphate-d34, 2-Ethyl-1-hexanol Hydrogen Phosphate A 208-d34, A 208 (phosphate)-d34, Baysolvex D 2EHPA-d34, Bis(2-ethylhexyl) Hydrogen Phosphate-d34, Bis(2-ethylhexyl) Orthophosphate-d34, Bis(2-ethylhexyl) phosphate-d34, D 2EHPA-d34, DEHAP-d34, DP 8-d34, DP 8R-d34, Di(2-ethylhexyl) Phosphate-d34, Di(2-ethylhexyl)orthophosphoric Acid-d34, Di(2-ethylhexyl)phosphoric Acid-d34, Di-2-ethylhexyl Hydrogen Phosphate-d34, Dioctyl Hydrogen Phosphate-d34, Dioctyl Orthophosphate-d34, Dioctyl Phosphate-d34, HDEHP-d34, HDEHPA-d34, Hostarex PA 216-d34, Hydrogen Bis(2-ethylhexyl) Phosphate-d34, JB 58-d34, LB 58-d34, O,O-Bis(2-ethylhexyl)phosphoric Acid-d34, O,O-Di(2-ethylhexyl)phosphoric Acid-d34, Orthophosphoric Acid 2-Ethylhexyl Alcohol Diester-d34, P 204-d34, P 204 (acid)-d34, Phoslex A 208-d34, Phosphoric Acid di(2-ethylhexyl)Ester-d34, TOPS 99-d34
InChI formel InChI=1S/C16H35O4P/c1-5-9-11-15(7-3)13-19-21(17,18)20-14-16(8-4)12-10-6-2/h15-16H,5-14H2,1-4H3,(H,17,18)/p-1/i1D3,2D3,3D3,4D3,5D2,6D2,7D2,8D2,9D2,10D2,11D2,12D2,13D2,14D2,15D,16D
SMIL [2H]C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])(C([2H])([2H])OP(=O)([O-])OC([2H])([2H])C([2H])(C([2H])([2H])C([2H])([2H])[2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])[2H])C([2H])([2H])C([2H])([2H])[2H]
IUPAC navn bis[1,1,2,3,3,4,4,5,5,6,6,6-dodecadeuterio-2-(1,1,2,2,2-pentadeuterioethyl)hexyl] phosphate
Molekylvægt (g/mol) 355.622
Formel vægt 355.433
Procent renhed >95
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.