Få mere at vide
Bis(2-ethylhexyl)maleate, TRC
Artikelnummer. 17931875
Skift visning
Mængde:
10 g
50 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17931875 50 g
17922675 10 g
17980835 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 17931875 Leverandør Toronto Research Chemicals Leverandørnr. TRCB43383550G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 142-16-5
Molekylær formel C20 H36 O4
Molekylvægt (g/mol) 340.5
Synonym Bis(2-ethylhexyl)maleate, 2-Butenedioic Acid (2Z)-Bis(2-ethylhexyl) Ester, 2-Butenedioic Acid (Z)-Bis(2-ethylhexyl) Ester, Maleic Acid Bis(2-ethylhexyl) Ester, Bis(2-ethylhexyl) Maleate, Ceraphyl 45, DOM, Di-2-ethylhexyl Maleate, Dioctyl Maleate
IUPAC navn bis(2-ethylhexyl) (Z)-but-2-enedioate
SMIL CCCCC(CC)COC(=O)\C=C/C(=O)OCC(CC)CCCC
Tekniske data
Kemisk navn eller materiale Bis(2 ethylhexyl)maleate
Mængde 50 g
InChI formel InChI=1S/C20H36O4/c1-5-9-11-17(7-3)15-23-19(21)13-14-20(22)24-16-18(8-4)12-10-6-2/h13-14,17-18H,5-12,15-16H2,1-4H3/b14-13-
Formel vægt 340.2614