Få mere at vide
Bis(2-ethylhexyl) Fumarate, TRC
Artikelnummer. 30133770
Skift visning
Mængde:
5 mL
10 mL
50 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30133770 50 mL
30136737 10 mL
30011859 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30133770 Leverandør Toronto Research Chemicals Leverandørnr. TRCB59203550ML
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 141-02-6
Molekylær formel C20H36O4
Molekylvægt (g/mol) 340.5
Synonym 2-Ethylhexyl fumarate, 2-Butenedioic acid (2E)-, 1,4-bis(2-ethylhexyl) ester (ACI), 2-Butenedioic acid (2E)-, bis(2-ethylhexyl) ester (9CI), 2-Butenedioic acid (E)-, bis(2-ethylhexyl) ester (ZCI), Fumaric acid, bis(2-ethylhexyl) ester (6CI, 7CI, 8CI), 2-Ethylhexyl fumarate, Bis(2-ethylhexyl) fumarate, Di-2-ethylhexyl fumarate, Dioctyl fumarate, DOF, Maximol FOK 670, RC Comonomer DOF
IUPAC navn bis(2-ethylhexyl) (E)-but-2-enedioate
SMIL CCCCC(CC)COC(=O)\C=C\C(=O)OCC(CC)CCCC
Tekniske data
Kemisk navn eller materiale Bis(2-ethylhexyl) Fumarate
Mængde 50 mL
InChI formel InChI=1S/C20H36O4/c1-5-9-11-17(7-3)15-23-19(21)13-14-20(22)24-16-18(8-4)12-10-6-2/h13-14,17-18H,5-12,15-16H2,1-4H3/b14-13+
Formel vægt 340.2614