Få mere at vide
Bis(2-chloroethyl)-d8 Ether, CDN
Artikelnummer. 30322356
Skift visning
Mængde:
0.05 g
0.1 g
Pakningsstørrelse:
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30322356 0.1 g 0.1g
30309866 0.05 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30322356 Leverandør CDN Leverandørnr. CDND24790.1G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Bis(2-chloroethyl)ether-d8
CAS 93952-02-4
Molekylær formel C4 D8 Cl2 O
Mængde 0.1 g
Synonym Ethane-1,1,2,2-d4, 1,1'-oxybis[2-chloro- (9 CI), Bis(2-chloroethyl-1,1,2,2-d4) Ether, Bis(2-chloroethyl) Ether-d8, Bis(2-chloroethyl) Ether-D8, 1,1'-Oxybis[2-chloroethane-1,1,2,2-d4], β,β'-Dichlorodiethyl ether-d8, β,β'-Dichlorodiethyl ether-d8
InChI formel InChI=1 S/C4H8Cl2O/c5-1-3-7-4-2-6/h1-4H2/i1D2,2D2,3D2,4D2
SMIL [2 H]C([2 H])(Cl)C([2 H])([2 H])OC([2 H])([2 H])C([2 H])([2 H])Cl
IUPAC navn 1-chloro-2-(2-chloro-1,1,2,2-tetradeuterioethoxy)-1,1,2,2-tetradeuterioethane
Molekylvægt (g/mol) 151.061
Formel vægt 150.0454 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity