Learn More
Bis(2-chloroethyl)amine Hydrochloride, TRC
Product Code. 30041057
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30041057 1 g 1g
30012033 5 g 5g
18090800 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Regulatory: This product fall under regulation “Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. If applicable, customers must provide a license before delivery.
Product Code. 30041057 Supplier Toronto Research Chemicals Supplier No. TRCB4188001G
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
Regulatory: This product fall under regulation “Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. If applicable, customers must provide a license before delivery.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 821-48-7
Molecular Formula C4 H9 Cl2 N . Cl H
Molecular Weight (g/mol) 178.49
Synonym Ethanamine, 2-chloro-N-(2-chloroethyl)-, hydrochloride (1:1), Diethylamine, 2,2'-dichloro-, hydrochloride (6CI,8CI), Ethanamine, 2-chloro-N-(2-chloroethyl)-, hydrochloride (9CI), 1,5-Dichloro-3-azapentane hydrochloride, 2,2'-Dichlorodiethylamine hydrochloride, 2-Chloro-N-(2-chloroethyl)ethanamine hydrochloride, Bi(2-chloroethyl)amine hydrochloride, Bis(2-chloroethyl)amine hydrochloride, Bis(2-chloroethyl)ammonium chloride, Bis(β-chloroethyl)amine hydrochloride, Di(2-chloroethyl)amine hydrochloride, Leo 72a, N,N-Bis(2-chloroethyl)amine hydrochloride, NSC 10873, Nornitrogen mustard hydrochloride, β,β'-Dichlorodiethylamine hydrochloride
IUPAC Name 2-chloro-N-(2-chloroethyl)ethanamine;hydrochloride
SMILES Cl.ClCCNCCCl
Specifications
Quantity 1 g
InChI Formula InChI=1S/C4H9Cl2N.ClH/c5-1-3-7-4-2-6;/h7H,1-4H2;1H
Formula Weight 176.9879
Percent Purity >95%
Chemical Name or Material Bis(2-chloroethyl)amine Hydrochloride
Grade HPLC