Få mere at vide
Bis(2-chloroethyl)amine-d4 Hydrochloride, TRC
Artikelnummer. 30345397
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30345397 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30345397 Leverandør Toronto Research Chemicals Leverandørnr. TRCB41880210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Bis(2-chloroethyl)amine-d4 Hydrochloride
CAS 58880-33-4
Molekylær formel C4H6D4Cl3N
Mængde 10 mg
Synonym Ethan-2,2-d2-amine, 2-chloro-N-(2-chloroethyl-2,2-d2)-, hydrochloride (9CI), Bis(2-chloroethyl)-2,2,2',2'-tetradeuterioamine hydrochloride, 2-Chloro-N-(2-chloroethyl-2,2-d2)ethan-2,2-d2-amine hydrochloride, Bis(2-chloroethyl)-2,2,2',2'-d4-amine Hydrochloride, Bis(2-chloroethyl)amine-D4 (bis(2-chloroethyl-2,2-D2)) Hydrochloride, Bis(2-chloroethyl)amine-2,2,2',2'-D4 Hydrochloride, Bis(2-chloroethyl)amine-d4 Hydrochloride
InChI formel InChI=1S/C4H9Cl2N.ClH/c5-1-3-7-4-2-6;/h7H,1-4H2;1H/i1D2,2D2;
SMIL Cl.[2H]C([2H])(Cl)CNCC([2H])([2H])Cl
IUPAC navn 2-chloro-N-(2-chloro-2,2-dideuterioethyl)-2,2-dideuterioethanamine;hydrochloride
Molekylvægt (g/mol) 182.51