Få mere at vide
Bis(2-chloroethyl)-1,1,2,2-d4-amine Hydrochloride, TRC
Artikelnummer. 30348070
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30348070 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30348070 Leverandør Toronto Research Chemicals Leverandørnr. TRCB41880310MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Bis(2-chloroethyl)-1,1,2,2-d4-amine hydrochloride
CAS 352431-06-2
Molekylær formel C4 2H4 H5 Cl2 N . Cl H
Mængde 10 mg
Synonym Ethan-1,1,2,2-d4-amine, 2-chloro-N-(2-chloroethyl)-, hydrochloride (9CI), Bis(2-chloroethyl)-1,1,2,2-tetradeuterioamine hydrochloride
InChI formel InChI=1S/C4H9Cl2N.ClH/c5-1-3-7-4-2-6;/h7H,1-4H2;1H/i1D2,3D2;
SMIL Cl.[2H]C([2H])(Cl)C([2H])([2H])NCCCl
IUPAC navn 2-chloro-N-(2-chloroethyl)-1,1,2,2-tetradeuterioethanamine;hydrochloride
Molekylvægt (g/mol) 182.51
Formel vægt 181.013