Få mere at vide
Bis(2-chloroethyl)-1,1,2,2-d4-amine HCl, CDN
Artikelnummer. 30325888
Skift visning
Mængde:
0.1 g
0.25 g
Pakningsstørrelse:
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30325888 0.25 g -
30310279 0.1 g 0.1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30325888 Leverandør CDN Leverandørnr. CDND53060.25G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Bis(2-chloroethyl)-1,1,2,2-d4-amine hydrochloride
CAS 352431-06-2
Molekylær formel C4 2H4 H5 Cl2 N . Cl H
Mængde 0.25 g
Synonym Ethan-1,1,2,2-d4-amine, 2-chloro-N-(2-chloroethyl)-, hydrochloride (9 CI), Bis(2-chloroethyl)-1,1,2,2-tetradeuterioamine hydrochloride
InChI formel InChI=1 S/C4H9Cl2N.ClH/c5-1-3-7-4-2-6;/h7H,1-4H2;1 H/i1D2,3D2;
SMIL Cl.[2 H]C([2 H])(Cl)C([2 H])([2 H])NCCCl
IUPAC navn 2-chloro-N-(2-chloroethyl)-1,1,2,2-tetradeuterioethanamine;hydrochloride
Molekylvægt (g/mol) 182.51
Formel vægt 181.013 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity