Få mere at vide
Bis(2-chloroethoxy)-d8-methane, CDN
Artikelnummer. 30327006
Skift visning
Mængde:
0.05 g
0.1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30327006 0.1 g
30309276 0.05 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30327006 Leverandør CDN Leverandørnr. CDND29390.1G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Bis(2-chloroethoxy)methane ethoxy-d8
CAS 93966-78-0
Molekylær formel C5 D8 H2 Cl2 O2
Mængde 0.1 g
Synonym Ethane-1,1,2,2-d4, 1,1'-[methylenebis(oxy)]bis[2-chloro- (9 CI), 1,1'-[Methylenebis(oxy)]bis[2-chloro-1,1,2,2-d4], 1,7-Dichloro-3,5-dioxaheptane-1,1,2,2,6,6,7,7-d8, 1-Chloro-2-(2-chloroethoxy-d4-methoxy)ethane-d4, Bis(2-chloroethoxy-d4)methane, Bis(2-chloroethoxy)-d8-methane, Bis(2-chloroethoxy)methane-D8 (bis-2-chloroethoxy-D8)
InChI formel InChI=1 S/C5H10Cl2O2/c6-1-3-8-5-9-4-2-7/h1-5H2/i1D2,2D2,3D2,4D2
SMIL [2 H]C([2 H])(Cl)C([2 H])([2 H])OCOC([2 H])([2 H])C([2 H])([2 H])Cl
IUPAC navn 1-chloro-2-[(2-chloro-1,1,2,2-tetradeuterioethoxy)methoxy]-1,1,2,2-tetradeuterioethane
Molekylvægt (g/mol) 181.087
Formel vægt 180.056 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity