missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Bis(2-butoxyethyl) Phthalate-d4, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Phthalic acid, bis-2-n-butoxyethyl ester D4 |
| CAS | 1398065-96-7 |
| Molekylær formel | C20 2H4 H26 O6 |
| Mængde | 25 mg |
| Synonym | Phthalic Acid, Bis-2-butoxyethyl Ester D4 (phenyl D4) |
| InChI formel | InChI=1S/C20H30O6/c1-3-5-11-23-13-15-25-19(21)17-9-7-8-10-18(17)20(22)26-16-14-24-12-6-4-2/h7-10H,3-6,11-16H2,1-2H3/i7D,8D,9D,10D |
| SMIL | [2H]c1c([2H])c([2H])c(C(=O)OCCOCCCC)c(C(=O)OCCOCCCC)c1[2H] |
| IUPAC navn | bis(2-butoxyethyl) 3,4,5,6-tetradeuteriobenzene-1,2-dicarboxylate |
| Molekylvægt (g/mol) | 370.47 |
| Formel vægt | 370.2293 |
Produkttitel
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?