missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bis(2-aminophenyl) ether, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
1 g
5 g
Description
It is catalytic reagent and used in pharmaceutical industry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is catalytic reagent and used in pharmaceutical industry.
Solubility
Slightly soluble in water.
Notes
Store at low temperature, away from light and moisture.
It is catalytic reagent and used in pharmaceutical industry.
Solubility
Slightly soluble in water.
Notes
Store at low temperature, away from light and moisture.
Specifications
Melting Point | 61°C to 64°C |
Color | Brown to White |
Quantity | 250 mg |
Solubility Information | Slightly soluble in water. |
Formula Weight | 200.24 |
Percent Purity | 98% |
Physical Form | Powder |
Chemical Name or Material | Bis(2-aminophenyl) ether |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'