missing translation for 'onlineSavingsMsg'
Learn More

Bis [2-[(2-Amino-1,6-dihydro-6-O-benzyl-9H-purin-9yl)methoxy]ethanol], TRC

Product Code. 30342438
Change view
Click to view available options
Mængde:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30342438 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30342438 Supplier Toronto Research Chemicals Supplier No. TRCB40180010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale Bis [2-[(2-Amino-1,6-dihydro-6-O-benzyl-9H-purin-9yl)methoxy]ethanol]
CAS 1346599-81-2
Molekylær formel C31H34N10O6
Mængde 10 mg
Synonym Bis [2-[(2-Amino-1,6-dihydro-6-phenylmethoxy-9H-purin-9yl)methoxy]ethanol]
InChI formel InChI=1S/C31H34N10O6/c42-11-13-44-20-40-18-34-24-26(40)36-30(38-28(24)46-15-22-7-3-1-4-8-22)32-17-33-31-37-27-25(35-19-41(27)21-45-14-12-43)29(39-31)47-16-23-9-5-2-6-10-23/h1-10,18-19,42-43H,11-17,20-21H2,(H,32,36,38)(H,33,37,39)
SMIL OCCOCn1cnc2c(OCc3ccccc3)nc(NCNc4nc(OCc5ccccc5)c6ncn(COCCO)c6n4)nc12
IUPAC navn 2-[[2-[[[9-(2-hydroxyethoxymethyl)-6-phenylmethoxypurin-2-yl]amino]methylamino]-6-phenylmethoxypurin-9-yl]methoxy]ethanol
Molekylvægt (g/mol) 642.67
Formel vægt 642.2663
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.