missing translation for 'onlineSavingsMsg'
Få mere at vide

Bis(2,2,2-Trichloroethyl) Phosphorochloridate, TRC

Product Code. 30341492
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Mængde:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30341492 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 30341492 missing translation for 'mfr' Toronto Research Chemicals missing translation for 'supplierNo' TRCB4950051G

Please to purchase this item. Need a web account? Register with us today!

Denne vare kræver en prisopdatering, inden den kan bestilles.

Anmod om en pris på denne vare her.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale Bis(2,2,2-Trichloroethyl) Phosphorochloridate
CAS 17672-53-6
Molekylær formel C4H4Cl7O3P
Mængde 1 g
Synonym Bis(2,2,2-trichloroethyl) Chlorophosphate, 1,1,1-Trichloro-2-[chloro(2,2,2-trichloroethoxy)phosphoryl]oxyethane
InChI formel InChI=1S/C4H4Cl7O3P/c5-3(6,7)1-13-15(11,12)14-2-4(8,9)10/h1-2H2
SMIL ClC(Cl)(Cl)COP(=O)(Cl)OCC(Cl)(Cl)Cl
IUPAC navn 1,1,1-trichloro-2-[chloro(2,2,2-trichloroethoxy)phosphoryl]oxyethane
Molekylvægt (g/mol) 379.22
Formel vægt 375.7718
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.