Få mere at vide
Bis(1-methylethyl) 2,6-Dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate, Mikromol™
Artikelnummer. 30052571
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30052571 25 mg
17912706 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30052571 Leverandør Mikromol Leverandørnr. MM0416.020025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Bis(1-methylethyl) 2,6-Dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Bis(1-methylethyl) 2,6-Dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate
CAS 21881-78-7
Analyt- eller komponentnavne Bis(1-methylethyl) 2,6-Dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate
Forsendelsestilstand Room Temperature
Molekylær formel C21 H26 N2 O6
Certificeringer/compliance ISO 17025
Synonym Bis(1-methylethyl) 2,6-Dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate, Nimodipine Imp. B (EP), Nimodipine Impurity B
InChI formel InChI=1S/C21H26N2O6/c1-11(2)28-20(24)17-13(5)22-14(6)18(21(25)29-12(3)4)19(17)15-8-7-9-16(10-15)23(26)27/h7-12,19,22H,1-6H3
SMIL CC(C)OC(=O)C1=C(C)NC(=C(C1c2cccc(c2)[N+](=O)[O-])C(=O)OC(C)C)C
IUPAC navn dipropan-2-yl 2,6-dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate
Molekylvægt (g/mol) 402.44
Mængde 25 mg
Formel vægt 402.1791 g/mol