Få mere at vide
Bis(1,3-dichloro-2-propyl) Phosphate, TRC
Artikelnummer. 30009633
Skift visning
Mængde:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30009633 100 mg
30007731 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30009633 Leverandør Toronto Research Chemicals Leverandørnr. TRCB419095100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 72236-72-7
Molekylær formel C6 H11 Cl4 O4 P
Molekylvægt (g/mol) 319.93
Synonym 2-Propanol, 1,3-dichloro-, hydrogen phosphate (9CI), Bis(1,3-dichloro-2-propyl) phosphate, 1,3-Dichloro-2-propanol Hydrogen Phosphate, 2-Propanol, 1,3-dichloro-, hydrogen phosphate (9CI)
IUPAC navn bis(1,3-dichloropropan-2-yl) hydrogen phosphate
SMIL OP(=O)(OC(CCl)CCl)OC(CCl)CCl
Tekniske data
Kemisk navn eller materiale Bis(1,3-dichloro-2-propyl) Phosphate
Renhedsgrad noter HPLC
Mængde 100 mg
InChI formel InChI=1S/C6H11Cl4O4P/c7-1-5(2-8)13-15(11,12)14-6(3-9)4-10/h5-6H,1-4H2,(H,11,12)
Formel vægt 317.9149
Procent renhed >95%