Få mere at vide
Biphenyl-d10, CDN
Artikelnummer. 30325408
Skift visning
Mængde:
1 g
Artikelnummer. Mængde
30325408 1 g
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30325408 Leverandør CDN Leverandørnr. CDND02081G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Biphenyl D10
CAS 1486-01-7
Molekylær formel C12 2H10
Mængde 1 g
Synonym 1,1'-Biphenyl-2,2',3,3',4,4',5,5',6,6'-d10, Biphenyl-d10 (6 CI,7 CI,8 CI), 1,1'-Biphenyl-d10, Perdeuteriobiphenyl, Perdeuterobiphenyl, Biphenyl D10
InChI formel InChI=1 S/C12H10/c1-3-7-11(8-4-1)12-9-5-2-6-10-12/h1-10 H/i1D,2 D,3 D,4 D,5 D,6 D,7 D,8 D,9 D,10 D
SMIL [2 H]c1c([2 H])c([2 H])c(c([2 H])c1[2 H])c2c([2 H])c([2 H])c([2 H])c([2 H])c2[2 H]
IUPAC navn 1,2,3,4,5-pentadeuterio-6-(2,3,4,5,6-pentadeuteriophenyl)benzene
Molekylvægt (g/mol) 164.27
Formel vægt 164.141 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity