missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Biphenyl-4-carboxylic acid, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
500 g
Description
Biphenyl-4-carboxylic acid is employed in the synthesis, characterization of europium, terbium complexes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Biphenyl-4-carboxylic acid is employed in the synthesis, characterization of europium, terbium complexes.
Solubility
Insoluble in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with bases and oxidizing agents.
Biphenyl-4-carboxylic acid is employed in the synthesis, characterization of europium, terbium complexes.
Solubility
Insoluble in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with bases and oxidizing agents.
Specifications
Melting Point | 223°C to 227°C |
Odor | Odorless |
Quantity | 25 g |
Beilstein | 973519 |
Solubility Information | Insoluble in water. |
Formula Weight | 198.22 |
Percent Purity | 98% |
Chemical Name or Material | Biphenyl-4-carboxylic acid |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'