Få mere at vide
Biphenamine hydrochloride 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Biphenamine Hydrochloride |
| CAS | 5560-62-3 |
| Analyt- eller komponentnavne | Biphenamine Hydrochloride |
| Forsendelsestilstand | +20°C |
| Molekylær formel | C19 H23 N O3 . Cl H |
| Koncentration | 100 μg/mL |
| Certificeringer/compliance | ISO 17034 |
| Synonym | [1,1'-Biphenyl]-3-carboxylic acid, 2-hydroxy-, 2-(diethylamino)ethyl ester, hydrochloride (1:1), 3-Biphenylcarboxylic acid, 2-hydroxy-, 2-(diethylamino)ethyl ester, hydrochloride (7CI,8CI), [1,1'-Biphenyl]-3-carboxylic acid, 2-hydroxy-, 2-(diethylamino)ethyl ester, hydrochloride (9CI), 2-(Diethylamino)ethyl 2-hydroxy-3-biphenylcarboxylate hydrochloride, 2-(Diethylamino)ethyl 3-Phenylsalicylate hydrochloride, Biphenamine hydrochloride, Melsaphine, Sebaclen, Sebaklen, Xenylsalate hydrochloride |
| InChI formel | InChI=1S/C19H23NO3.ClH/c1-3-20(4-2)13-14-23-19(22)17-12-8-11-16(18(17)21)15-9-6-5-7-10-15;/h5-12,21H,3-4,13-14H2,1-2H3;1H |
| SMIL | Cl.CCN(CC)CCOC(=O)c1cccc(c1O)c2ccccc2 |
| Vis mere |