missing translation for 'onlineSavingsMsg'
Få mere at vide

Biperiden hydrochloride, Tocris Bioscience™

Artikelnummer. 18782754
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
18782754 50 mg
18702714 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Regulatorisk: Dette produkt falder ind under Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. Hvis det er relevant, skal kunden fremskaffe en licens før levering.
Artikelnummer. 18782754 Leverandør Tocris Bioscience Leverandørnr. 2853/50

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter

Denne vare kan ikke returneres. Se returpolicy
Regulatorisk: Dette produkt falder ind under Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. Hvis det er relevant, skal kunden fremskaffe en licens før levering.

Muscarinic receptor antagonist; some selectivity for M1

Muscarinic receptor antagonist that displays some selectivity for the M1 subtype (Ki values are 0.48, 2.4, 3.9, 6.3 and 6.3 nM for M1, M4, M3, M2 and M5 receptors respectively). Alleviates extrapyramidal symptoms associated with antipsychotic drugs. Displays antiparkinsonian activity.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1235-82-1
Molekylær formel C21H30ClNO
Molekylvægt (g/mol) 347.927
InChI nøgle RDNLAULGBSQZMP-LIJKWCKLSA-N
Synonym 1r-1-1r,4r,5s-5-bicyclo 2.2.1 hept-2-enyl-1-phenyl-3-piperidin-1-ylpropan-1-ol;hydrochloride
PubChem CID 129318274
IUPAC navn (1R)-1-[(1R,4R,5S)-5-bicyclo[2.2.1]hept-2-enyl]-1-phenyl-3-piperidin-1-ylpropan-1-ol;hydrochloride
SMIL C1CCN(CC1)CCC(C2CC3CC2C=C3)(C4=CC=CC=C4)O.Cl

Specifications

Kemisk navn eller materiale Biperiden hydrochloride
Mængde 50 mg
Formel vægt 347.92
Procent renhed >99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.